Alendronate sodium ≥98%

Supplier: Matrix Scientific

Synonyms: Alendronate monosodium

047878-1G 047878-5G
101881-818EA 93.85 USD
101881-818 101881-820
Alendronate sodium ≥98%
Alendronate sodium

Hazard: Irritant

Formula: C₄H₁₂NNaO₇P₂
MW: 271.08 g/mol
MDL Number: MFCD01861681
CAS Number: 129318-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR