2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥95%
Supplier: Matrix Scientific
047876-500MG
047876-1G
047876-5G
101881-806EA
181.54
USD
101881-806
101881-808
101881-810
2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥95%
2-(2-Methoxyphenoxy)ethanamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₄ClNO₂ |
MDL Number:
MFCD01631120 CAS Number: 64464-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...