4-Glycidyloxycarbazole ≥96%

Supplier: Matrix Scientific

Synonyms: 4-(2,3-Epoxypropoxy)carbazole, Carvedilol Related Compound D

047874-1G 047874-5G
101881-796EA 192.31 USD
101881-796 101881-798
4-Glycidyloxycarbazole ≥96%
4-Glycidyloxycarbazole

Hazard: Irritant

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
MDL Number: MFCD03411884
CAS Number: 51997-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR