Methyl-5-(2,4-dichlorophenyl)-4-(hydroxymethyl)-1,2-dimethyl-2-pyrrolidinecarboxylate ≥95%

Supplier: Matrix Scientific

047872-500MG 047872-1G
101881-786EA 921.54 USD
101881-786 101881-788
Methyl-5-(2,4-dichlorophenyl)-4-(hydroxymethyl)-1,2-dimethyl-2-pyrrolidinecarboxylate ≥95%
Methyl-5-(2,4-dichlorophenyl)-4-(hydroxymethyl)-1,2-dimethyl-2-pyrrolidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₉Cl₂NO₃
MDL Number: MFCD09027171
CAS Number: 1052611-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR