[4-(Aminomethyl)-1,2-dimethyl-5-phenyl-2-pyrrolidinyl]methanol ≥95%

Supplier: Matrix Scientific

047871-500MG 047871-1G
101881-782EA 921.54 USD
101881-782 101881-784
[4-(Aminomethyl)-1,2-dimethyl-5-phenyl-2-pyrrolidinyl]methanol ≥95%
[4-(Aminomethyl)-1,2-dimethyl-5-phenyl-2-pyrrolidinyl]methanol

Hazard: Irritant

Formula: C₁₄H₂₂N₂O
MDL Number: MFCD09027170
CAS Number: 1052611-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR