5-(Methoxycarbonyl)-5-methyl-2-(3-pyridinyl)-3-pyrrolidinecarboxylic acid dihydrochloride ≥95%

Supplier: Matrix Scientific

047867-500MG 047867-1G
101881-766EA 921.54 USD
101881-766 101881-768
5-(Methoxycarbonyl)-5-methyl-2-(3-pyridinyl)-3-pyrrolidinecarboxylic acid dihydrochloride ≥95%
5-(Methoxycarbonyl)-5-methyl-2-(3-pyridinyl)-3-pyrrolidinecarboxylic acid dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₄
MDL Number: MFCD09027166
CAS Number: 1177765-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR