5-(Methoxycarbonyl)-1,5-dimethyl-2-[2-(trifluoromethyl)phenyl]-3-pyrrolidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047866-500MG 047866-1G
101881-762EA 921.54 USD
101881-762 101881-764
5-(Methoxycarbonyl)-1,5-dimethyl-2-[2-(trifluoromethyl)phenyl]-3-pyrrolidinecarboxylic acid ≥95%
5-(Methoxycarbonyl)-1,5-dimethyl-2-[2-(trifluoromethyl)phenyl]-3-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₈F₃NO₄
MW: 345.32 g/mol
MDL Number: MFCD08689808
CAS Number: 1052611-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR