2-(2,3-Difluorophenyl)-5-(methoxycarbonyl)-1-methyl-3-pyrrolidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

047859-500MG 047859-1G
101881-734EA 921.54 USD
101881-734 101881-736
2-(2,3-Difluorophenyl)-5-(methoxycarbonyl)-1-methyl-3-pyrrolidinecarboxylic acid ≥95%
2-(2,3-Difluorophenyl)-5-(methoxycarbonyl)-1-methyl-3-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₅F₂NO₄
MDL Number: MFCD08064793
CAS Number: 1096442-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR