1-(tert-Butyl)-5-methyl-3-(2-aminoethyl)-1H-indole-1,5-dicarboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

047853-500MG 047853-1G
101881-710EA 1296.92 USD
101881-710 101881-712
1-(tert-Butyl)-5-methyl-3-(2-aminoethyl)-1H-indole-1,5-dicarboxylate hydrochloride ≥95%
1-(tert-Butyl)-5-methyl-3-(2-aminoethyl)-1H-indole-1,5-dicarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₃ClN₂O₄
MDL Number: MFCD07698598
CAS Number: 1171456-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR