3-Chloro-5-(trifluoromethyl)-2-pyridinyl-4-piperidinyl ether ≥95%
Supplier: Matrix Scientific
047840-500MG
047840-1G
101881-664EA
747.69
USD
101881-664
101881-666
3-Chloro-5-(trifluoromethyl)-2-pyridinyl-4-piperidinyl ether ≥95%
3-Chloro-5-(trifluoromethyl)-2-pyridinyl-4-piperidinyl ether
Hazard: Irritant
Formula:
C₁₁H₁₂ClF₃N₂O MW: 280.68 g/mol |
MDL Number:
MFCD11553058 CAS Number: 1092352-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...