tert-Butyl-4-[(2-ethoxy-2-oxoethyl)amino]tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

047832-500MG 047832-1G
101881-632EA 391 USD
101881-632 101881-634
tert-Butyl-4-[(2-ethoxy-2-oxoethyl)amino]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-[(2-ethoxy-2-oxoethyl)amino]tetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₄
MW: 286.37 g/mol
MDL Number: MFCD11046697
CAS Number: 177276-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR