Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-fluoro-2-(trifluoromethyl)benzoate, 4-Fluoro-2-trifluoromethylbenzoic acid methyl ester
047830-500MG
047830-1G
047830-5G
101881-622EA
153.85
USD
101881-622
101881-624
101881-626
Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate ≥95%
Methyl-4-fluoro-2-(trifluoromethyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₉H₆F₄O₂ MW: 222.14 g/mol |
MDL Number:
MFCD06203708 CAS Number: 220141-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...