Methyl-5-bromo-2-{[(ethylamino)carbonyl]amino}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047813-500MG 047813-1G 047813-5G
101881-558EA 278.46 USD
101881-558 101881-560 101881-562
Methyl-5-bromo-2-{[(ethylamino)carbonyl]amino}benzenecarboxylate ≥95%
Methyl-5-bromo-2-{[(ethylamino)carbonyl]amino}benzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂O₃
MDL Number: MFCD03145717
CAS Number: 433253-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR