N-(2-Hydroxyethyl)-N-methylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

047799-500MG 047799-1G 047799-5G
101881-492EA 181.54 USD
101881-492 101881-494 101881-496
N-(2-Hydroxyethyl)-N-methylbenzenesulfonamide ≥95%
N-(2-Hydroxyethyl)-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₃NO₃S
MDL Number: MFCD00591616
CAS Number: 59724-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR