4-Amino-3-fluorothiophenol ≥95%

Supplier: Matrix Scientific

047796-500MG 047796-1G 047796-5G
101881-478EA 278.46 USD
101881-478 101881-480 101881-482
4-Amino-3-fluorothiophenol ≥95%
4-Amino-3-fluorothiophenol

Hazard: Irritant/Light Sensitive/Refrigerate/Stored under Argon

Formula: C₆H₆FNS
MDL Number: MFCD09835280
CAS Number: 15178-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR