Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-{[4-(trifluoromethyl)piperidin-1-yl]methyl}benzoate

047791-500MG 047791-1G
101881-452EA 375.38 USD
101881-452 101881-454
Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate ≥95%
Methyl-3-{[4-(trifluoromethyl)piperidino]methyl}benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₂
MW: 301.31 g/mol
MDL Number: MFCD09864681
CAS Number: 1000339-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR