3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%

Supplier: Matrix Scientific

047784-500MG 047784-1G 047784-5G
101881-422EA 181.54 USD
101881-422 101881-424 101881-426
3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide ≥95%
3-(4-Bromophenyl)-5,6-dihydroimidazo[2,1-b][1,3]thiazole hydrobromide

Hazard: Irritant

Formula: C₁₁H₁₀Br₂N₂S
MDL Number: MFCD00816059
CAS Number: 36104-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR