tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(piperazin-1-yl)piperidine-1-carboxylate, 4-(Piperazin-1-yl)piperidine, N-BOC protected
047764-500MG
047764-1G
047764-5G
101881-312EA
424.62
USD
101881-312
101881-314
101881-316
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-piperazinotetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₂₇N₃O₂ MW: 269.39 g/mol |
MDL Number:
MFCD04973999 CAS Number: 177276-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...