Ethyl-2-(benzylamino)-2-(hydroxyimino)acetate ≥95%

Supplier: Matrix Scientific

047757-500MG 047757-1G 047757-5G
101881-278EA 278.46 USD
101881-278 101881-280 101881-282
Ethyl-2-(benzylamino)-2-(hydroxyimino)acetate ≥95%
Ethyl-2-(benzylamino)-2-(hydroxyimino)acetate

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD09027151
CAS Number: 937604-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR