2-Fluoro-5-nitrophenol ≥95%

Supplier: Matrix Scientific

046618-5G 046618-25G
101876-300EA 106.15 USD
101876-300 101876-302
2-Fluoro-5-nitrophenol ≥95%
2-Fluoro-5-nitrophenol

Hazard: Irritant

Formula: C₆H₄FNO₃
MW: 157.1000 g/mol
MDL Number: MFCD09054718
CAS Number: 22510-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR