Methyl-2-(4-nitrophenyl)butanoate ≥95%

Supplier: Matrix Scientific

046617-500MG 046617-1G 046617-5G
101876-294EA 181.54 USD
101876-294 101876-296 101876-298
Methyl-2-(4-nitrophenyl)butanoate ≥95%
Methyl-2-(4-nitrophenyl)butanoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD10758074
CAS Number: 24646-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR