4-Chloro-2-[(dimethylamino)methyl]-6-methylbenzenol ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-2-[(dimethylamino)methyl]-6-methylphenol
046531-500MG
046531-1G
046531-5G
101875-922EA
278.46
USD
101875-922
101875-924
101875-926
4-Chloro-2-[(dimethylamino)methyl]-6-methylbenzenol ≥95%
4-Chloro-2-[(dimethylamino)methyl]-6-methylbenzenol
Hazard: Irritant
Formula:
C₁₀H₁₄ClNO |
MDL Number:
MFCD08443991 CAS Number: 87117-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...