2-Bromo-4,6-dimethylbenzenol ≥95%

Supplier: Matrix Scientific

046510-1G 046510-5G
101875-812EA 272.31 USD
101875-812 101875-814
2-Bromo-4,6-dimethylbenzenol ≥95%
2-Bromo-4,6-dimethylbenzenol

Hazard: Irritant

Formula: C₈H₉BrO
MDL Number: MFCD08443978
CAS Number: 15191-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR