2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)phenol

046509-500MG 046509-1G 046509-5G
101875-804EA 278.46 USD
101875-804 101875-806 101875-808
2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol ≥95%
2,6-Bis[(dimethylamino)methyl]-4-(2-methylbutyl)benzenol

Hazard: Irritant

Formula: C₁₇H₃₀N₂O
MW: 278.44 g/mol
MDL Number: MFCD08443977
CAS Number: 924868-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR