2-[(Dimethylamino)methyl]-4,6-dimethylbenzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dimethyl-6-[(dimethylamino)methyl]phenol

046508-500MG 046508-1G 046508-5G
101875-798EA 278.46 USD
101875-798 101875-800 101875-802
2-[(Dimethylamino)methyl]-4,6-dimethylbenzenol ≥95%
2-[(Dimethylamino)methyl]-4,6-dimethylbenzenol

Hazard: Irritant

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
MDL Number: MFCD08443976
CAS Number: 52777-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR