2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Bis[(dimethylamino)methyl]-4-pent-2-ylphenol

046507-500MG 046507-1G 046507-5G
101875-792EA 278.46 USD
101875-792 101875-794 101875-796
2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol ≥95%
2,6-Bis[(dimethylamino)methyl]-4-(1-methylbutyl)benzenol

Hazard: Irritant

Formula: C₁₇H₃₀N₂O
MW: 278.44 g/mol
MDL Number: MFCD08443975
CAS Number: 924868-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR