2-[(Dimethylamino)methyl]-4-ethylbenzenol ≥90%
Supplier: Matrix Scientific
046505-500MG
046505-1G
046505-5G
101875-780EA
278.46
USD
101875-780
101875-782
101875-784
2-[(Dimethylamino)methyl]-4-ethylbenzenol ≥90%
2-[(Dimethylamino)methyl]-4-ethylbenzenol
Hazard: Irritant
Formula:
C₁₁H₁₇NO MW: 179.26 g/mol |
MDL Number:
MFCD01734031 CAS Number: 55955-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...