Ethyl-1-[1-methyl-2-oxo-2-(2-thienyl)ethyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

046502-500MG 046502-1G 046502-5G
101875-768EA 375.38 USD
101875-768 101875-770 101875-772
Ethyl-1-[1-methyl-2-oxo-2-(2-thienyl)ethyl]-4-piperidinecarboxylate ≥95%
Ethyl-1-[1-methyl-2-oxo-2-(2-thienyl)ethyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₂₁NO₃S
MDL Number: MFCD08443973
CAS Number: 924868-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR