2-Bromo-1-(2-thienyl)-1-propanone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-1-(thiophen-2-yl)propan-1-one
046499-500MG
046499-1G
046499-5G
101875-750EA
181.54
USD
101875-750
101875-752
101875-754
2-Bromo-1-(2-thienyl)-1-propanone ≥95%
2-Bromo-1-(2-thienyl)-1-propanone
Hazard: Irritant
Formula:
C₇H₇BrOS MW: 219.0989 g/mol |
MDL Number:
MFCD08443970 CAS Number: 75815-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...