Methyl-2-bromo-6-(trifluoromethyl)nicotinate ≥95%
Supplier: Matrix Scientific
046474-500MG
046474-1G
101875-636EA
556.92
USD
101875-636
101875-638
Methyl-2-bromo-6-(trifluoromethyl)nicotinate ≥95%
Methyl-2-bromo-6-(trifluoromethyl)nicotinate
Hazard: Irritant
Formula:
C₈H₅BrF₃NO₂ MW: 284.04 g/mol |
MDL Number:
MFCD12025861 CAS Number: 144740-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...