Potassium-5-cyano-1,1,1-trifluoro-6-methoxy-6-oxo-2,4-hexadien-2-olate ≥95%

Supplier: Matrix Scientific

046473-500MG 046473-1G 046473-5G
101875-630EA 278.46 USD
101875-630 101875-632 101875-634
Potassium-5-cyano-1,1,1-trifluoro-6-methoxy-6-oxo-2,4-hexadien-2-olate ≥95%
Potassium-5-cyano-1,1,1-trifluoro-6-methoxy-6-oxo-2,4-hexadien-2-olate

Hazard: Irritant

Formula: C₈H₅F₃KNO₃
MDL Number: MFCD12025860
CAS Number: 1001924-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR