Ethyl-1-methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylate ≥95%

Supplier: Matrix Scientific

046472-500MG 046472-1G
101875-626EA 556.92 USD
101875-626 101875-628
Ethyl-1-methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylate ≥95%
Ethyl-1-methyl-4-(trifluoromethyl)-1H-pyrrole-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₀F₃NO₂
MDL Number: MFCD12025859
CAS Number: 130511-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR