Diethyl-4-chloro-3,5-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

046462-500MG 046462-1G
101875-594EA 375.38 USD
101875-594 101875-596
Diethyl-4-chloro-3,5-pyridinedicarboxylate ≥95%
Diethyl-4-chloro-3,5-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₄
MW: 257.68 g/mol
MDL Number: MFCD12025855
CAS Number: 244638-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR