Diethyl-4-oxo-1,4-dihydro-3,5-pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

046460-500MG 046460-1G
101875-584EA 278.46 USD
101875-584 101875-586
Diethyl-4-oxo-1,4-dihydro-3,5-pyridinedicarboxylate ≥95%
Diethyl-4-oxo-1,4-dihydro-3,5-pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₅
MDL Number: MFCD12025854
CAS Number: 74632-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR