Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate ≥95%
Supplier: Matrix Scientific
046455-500MG
046455-1G
101875-566EA
375.38
USD
101875-566
101875-568
Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate ≥95%
Methyl-6-[4-(trifluoromethyl)piperidino]nicotinate
Hazard: Irritant
Formula:
C₁₃H₁₅F₃N₂O₂ |
MDL Number:
MFCD10758065 CAS Number: 1033463-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...