2,4-Dichloro-3-formylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,4-Dichloronicotinaldehyde, 2,4-Dichloropyridine-3-carboxaldehyde, 2,4-Dichloro-3-formylpyridine, 2,4-Dichloro-3-pyridinecarboxaldehyde
046449-1G
046449-5G
101875-538EA
226.15
USD
101875-538
101875-540
2,4-Dichloro-3-formylpyridine ≥95%
2,4-Dichloro-3-formylpyridine
Hazard: Irritant/Refrigerate
Formula:
C₆H₃Cl₂NO MW: 176.00 g/mol |
MDL Number:
MFCD07437909 CAS Number: 134031-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...