1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine ≥95%

Supplier: Matrix Scientific

046442-500MG 046442-1G
101875-514EA 278.46 USD
101875-514 101875-516
1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine ≥95%
1-(5-Nitro-2-pyridinyl)-4-(trifluoromethyl)piperidine

Hazard: Irritant

Formula: C₁₁H₁₂F₃N₃O₂
MDL Number: MFCD00204721
CAS Number: 175277-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR