5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046436-500MG 046436-1G
101875-484EA 747.69 USD
101875-484 101875-486
5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Chloro-1-(4-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₉ClFNO₂
MW: 265.67 g/mol
MDL Number: MFCD10758060
CAS Number: 1033463-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR