5-Chloro-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046434-500MG 046434-1G
101875-476EA 747.69 USD
101875-476 101875-478
5-Chloro-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Chloro-1-(2-chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈Cl₂FNO₂
MDL Number: MFCD10758058
CAS Number: 1033463-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR