2-(Pyrrolidin-3-yl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Pyridin-2-yl)pyrrolidine

046425-500MG 046425-1G
101875-440EA 321.54 USD
101875-440 101875-442
2-(Pyrrolidin-3-yl)pyridine ≥95%
2-(Pyrrolidin-3-yl)pyridine

Hazard: Irritant

Formula: C₉H₁₂N₂
MW: 148.20 g/mol
MDL Number: MFCD01862539
CAS Number: 150281-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR