2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
046406-500MG
046406-1G
046406-5G
101875-362EA
375.38
USD
101875-362
101875-364
101875-366
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide ≥95%
2-Chloro-4-(1H-1,2,4-triazol-1-yl)benzenecarbohydrazide
Hazard: Irritant
Formula:
C₉H₈ClN₅O MW: 237.65 g/mol |
MDL Number:
MFCD09607898 CAS Number: 952183-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...