Ethyl-2-methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoate ≥95%

Supplier: Matrix Scientific

046398-500MG 046398-1G 046398-5G
101875-326EA 375.38 USD
101875-326 101875-328 101875-330
Ethyl-2-methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoate ≥95%
Ethyl-2-methyl-2-[(5-nitro-2-pyridinyl)sulfanyl]propanoate

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄S
MW: 270.31 g/mol
MDL Number: MFCD09152777
CAS Number: 952183-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR