1-Benzyl-2-(tert-butyl)-1,2-diazepane-1,2-dicarboxylate ≥95%

Supplier: Matrix Scientific

046375-500MG 046375-1G 046375-5G
101875-224EA 375.38 USD
101875-224 101875-226 101875-228
1-Benzyl-2-(tert-butyl)-1,2-diazepane-1,2-dicarboxylate ≥95%
1-Benzyl-2-(tert-butyl)-1,2-diazepane-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₄
MDL Number: MFCD11840990
CAS Number: 443295-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR