4-(sec-Butyl)-2-[(dimethylamino)methyl]benzenol ≥90%
Supplier: Matrix Scientific
046361-500MG
046361-1G
046361-5G
101875-160EA
278.46
USD
101875-160
101875-162
101875-164
4-(sec-Butyl)-2-[(dimethylamino)methyl]benzenol ≥90%
4-(sec-Butyl)-2-[(dimethylamino)methyl]benzenol
Hazard: Irritant
Formula:
C₁₃H₂₁NO |
MDL Number:
MFCD11100201 CAS Number: 92111-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...