2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol ≥95%

Supplier: Matrix Scientific

046360-500MG 046360-1G 046360-5G
101875-154EA 278.46 USD
101875-154 101875-156 101875-158
2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol ≥95%
2,6-Bis[(dimethylamino)methyl]-4-ethylbenzenol

Hazard: Irritant

Formula: C₁₄H₂₄N₂O
MDL Number: MFCD08443957
CAS Number: 36805-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR