4-Chloro-6-piperidino-5-pyrimidinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046348-500MG 046348-1G 046348-5G
101875-094EA 375.38 USD
101875-094 101875-096 101875-098
4-Chloro-6-piperidino-5-pyrimidinecarbaldehyde ≥95%
4-Chloro-6-piperidino-5-pyrimidinecarbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MW: 225.68 g/mol
MDL Number: MFCD08443953
CAS Number: 54503-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR