Methyl (4-bromophenyl)acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
methyl 2-(4-bromophenyl)acetate
046346-5G
046346-25G
101875-088EA
30.77
USD
101875-088
101875-090
Methyl (4-bromophenyl)acetate ≥95%
Methyl (4-bromophenyl)acetate
Hazard: Irritant
Formula:
C₉H₉BrO₂ MW: 229.0700 g/mol |
MDL Number:
MFCD00126844 CAS Number: 41841-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...