4-tert-Butylbenzamide ≥95%

Supplier: Matrix Scientific

046298-5G 046298-25G
101874-986EA 86.15 USD
101874-986 101874-988
4-tert-Butylbenzamide ≥95%
4-tert-Butylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD00017139
CAS Number: 56108-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR