2-Bromo-3'-chloropropiophenone ≥95%

Supplier: Matrix Scientific

046293-5G 046293-25G 046293-100G
101874-972EA 44.62 USD
101874-972 101874-974 101874-976
2-Bromo-3'-chloropropiophenone ≥95%
2-Bromo-3'-chloropropiophenone

Hazard: Irritant

Formula: C₉H₈BrClO
MDL Number: MFCD03701150
CAS Number: 34911-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR