5-Chloro-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

046287-500MG 046287-1G
101874-958EA 692.31 USD
101874-958 101874-960
5-Chloro-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%
5-Chloro-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Hazard: Irritant

Formula: C₉H₉ClN₄
MDL Number: MFCD12025853
CAS Number: 1135283-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR